missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GEMIN2 Monoclonal Antibody (OTI6G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808823S
This item is not returnable.
View return policy
Description
GEMIN2 Monoclonal antibody specifically detects GEMIN2 in Human samples. It is validated for Western BlotSpecifications
| GEMIN2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8NEJ9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 25983 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6G4 | |
| Unconjugated | |
| Ngdn | |
| 1700012N19Rik; Component of gems 2; gem (nuclear organelle) associated protein 2; gem nuclear organelle associated protein 2; gem nuclear organelle associated protein 2 S homeolog; gem-associated protein 2; GEMIN2; Gemin-2; gemin2.S; SIP 1; SIP1; SIP-1; SIP1 delta; sip1-delta; SMN interacting protein 1-delta; SMN interacting protein-1; SMN-interacting protein 1; survival of motor neuron protein interacting protein 1; Survival of motor neuron protein-interacting protein 1; survivor of motor neuron protein interacting protein 1; XELAEV_18042638mg | |
| Full length human recombinant protein of human NGDN produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction