Learn More
GEMIN8 Antibody (OTI1E5), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71977AF700
Product Code. 18112865
  • € 751.00 / 0.10mL
Description
GEMIN8 Monoclonal specifically detects GEMIN8 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GEMIN8
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
GEMIN8
Full length recombinant protein of human GEMIN8 (NP_060326) produced in E.coli.
0.1 mL
54960.0
Store at 4°C in the dark.
Western Blot, Immunohistochemistry
OTI1E5
Western Blot, Immunohistochemistry
FAM51A1, FLJ43072, gem (nuclear organelle) associated protein 8, Gemin-8, member A1, Protein FAM51A1
Mouse
Affinity Purified