missing translation for 'onlineSavingsMsg'
Learn More

Geminin Antibody (CPTC-GMNN-1), CoraFluor™ 1, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-79878CL1

Product Code. 30151849

  • € 1165.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Geminin Monoclonal antibody specifically detects Geminin in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Geminin
Monoclonal
CoraFluor 1
Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3
Recombinant human full-length protein (Uniprot: O75496)
0.1 mL
Breast Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Receptors Coactivators and Corepressors, Stem Cell Markers
51053
Store at 4°C in the dark. Do not freeze.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array, Immunocytochemistry/Immunofluorescence
CPTC-GMNN-1
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.