missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gemtuzumab Antibody (975155) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB96171C
This item is not returnable.
View return policy
Description
Gemtuzumab Monoclonal antibody specifically detects Gemtuzumab in Broad Species Reactivity samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| Gemtuzumab | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Full length Gemtuzumab | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA | |
| 975155 | |
| Western Blot, Flow Cytometry, ELISA | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction