Learn More
Invitrogen™ GEN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587557
Description
Immunogen sequence: LQDLPLKERI FTKLSYPQDN LQPDVNLKTL SILSVKESCI ANSGSDCTSH LSKDLPGIPL QNESRDSKIL KGDQLLQEDY KVNTSVPYSV SNTVVKTCNV RPPNTALDHS RKVDMQTTRK ILMKKSVCLD RHSSDEQSAP VFGKAKYTTQ RMKHSSQKHN SSHFKESGHN KLSSPKIHIK ETEQCVRSYE TAENEESCFP DSTKSSLSSL QCHKKENNSG TCLDSPLPLR QRLKLRFQST; Positive Samples: H460, MCF-7, Mouse thymus, Mouse spleen; Cellular Location: Nucleus.
Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.
Specifications
| GEN1 | |
| Polyclonal | |
| Unconjugated | |
| GEN1 | |
| 5830483C08Rik; Flap endonuclease GEN homolog 1; Gen; Gen endonuclease homolog 1; Gen homolog 1, endonuclease; Gen homolog 1, endonuclease (Drosophila); GEN1; GEN1 Holliday junction 5' flap endonuclease; GEN1, Holliday junction 5' flap endonuclease; Holliday junction resolvase; RGD1559792 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 209334, 348654 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q17RS7, Q8BMI4 | |
| GEN1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 669-908 of human GEN1 (NP_8724313). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.