Learn More
Kartell™ General Purpose Funnels
Tough, robust, general purpose funnels suitable for a wide range of liquids
Brand: Kartell™ 0417900
Additional Details : Weight : 5.77000kg
Description
- Upstand on rim to reduce possible overflow
- Ribbed on outside to eliminate back pressure
- Hanging eye for storage
- High-density polyethylene (HDPE)
- Suitable for a wide range of liquids
Specifications
420 mm | |
HDPE | |
138 mm |
10,000 mL | |
57 mm |