Learn More
Buerkle™ General Purpose Aseptic Polypropylene Sample Box
Buerkle™ General Purpose Aseptic Polypropylene Sample Box features a lit that can be opened with one hand and a 90° angle hinge.
Brand:  Buerkle™ 6206-0100
.55 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 4.90000kg
 View more versions of this product
Product Code. 15723209
  • € 0.60 / Each
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
Description
Description
  • PP, transparent
  • With area for labelling and with graduation
  • Liquid/condensate in the lid runs into an absorbing groove integrated in the lid, nothing is lost, nothing can drip
  • The box is completely tight, has no crevices, is therefore easy to fill and can be emptied without residues
  • Produced under aseptic conditions
  • Autoclavable and suitable for deep freezing
Specifications
Specifications
Sample Box
43 mm
Transparent
90 mL
Suitable for Use with Foodstuffs
1
Polypropylene
Sealing Cover with Absorbing Groove
Translucent
Aseptic Sample box
Autoclavable
Polypropylene
Sealing cover
Yes
80 mm