Learn More
Welch™ Generator Atmosphere
Shop All Welch Ilmvac Products
Click to view available options
Pump Type:
Generator Atmosphere
Unit Size:
Each
Specifications
Specifications
| Motor Speed | 13 to 15 mL/min. |
| Noise Level | 44 dB(A) |
| Frequency | 50/60 Hz |
| Pump Type | Generator Atmosphere |