Learn More
Gentiopicroside, MedChemExpress
Product Code. 16468998
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
Product Code. 16468998
Brand: Medchem Express HYN0494
MedChemExpress Gentiopicroside, a naturally occurring iridoid glycoside, inhibits P450 activity, with an IC50 and a Ki of 61 μM and 22.8 μM for CYP2A6; Gentiopicroside has antianti-inflammatoryand antioxidative effects.
Gentiopicroside, a naturally occurring iridoid glycoside, inhibits P450 activity, with an IC50 and a Ki of 61 μM and 22.8 μM for CYP2A6
Specifications
Chemical Name or Material Gentiopicroside
CAS 20831-76-9
Color White
Purity Grade Notes Research
Molecular Formula C16H20O9
Quantity 10 mg
Synonym Gentiopicrin
Solubility Information DMSO : ≥ 100 mg/mL (280.65 mM) ∣H2O : 100 mg/mL (280.65 mM; Need ultrasonic)
SMILES C=C[C@@H]([C@@H]1O[C@]([C@@H]([C@@H](O)[C@@H]2O)O)([H])O[C@@H]2CO)C(C3=CO1)=CCOC3=O
Molecular Weight (g/mol) 356.32
Formula Weight 356.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only