missing translation for 'onlineSavingsMsg'
Learn More

Gentiopicroside, MedChemExpress

Product Code. 16439096
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16439096

Brand: Medchem Express HYN0494

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gentiopicroside, a naturally occurring iridoid glycoside, inhibits P450 activity, with an IC50 and a Ki of 61 μM and 22.8 μM for CYP2A6; Gentiopicroside has antianti-inflammatoryand antioxidative effects.

Gentiopicroside, a naturally occurring iridoid glycoside, inhibits P450 activity, with an IC50 and a Ki of 61 μM and 22.8 μM for CYP2A6

Specifications

Chemical Name or Material Gentiopicroside
CAS 20831-76-9
Color White
Purity Grade Notes Research
Molecular Formula C16H20O9
Quantity 100 mg
Synonym Gentiopicrin
Solubility Information DMSO : ≥ 100 mg/mL (280.65 mM) ∣H2O : 100 mg/mL (280.65 mM; Need ultrasonic)
SMILES C=C[C@@H]([C@@H]1O[C@]([C@@H]([C@@H](O)[C@@H]2O)O)([H])O[C@@H]2CO)C(C3=CO1)=CCOC3=O
Molecular Weight (g/mol) 356.32
Formula Weight 356.32
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.