missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gephyrin/GPHN Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-34859CL1
This item is not returnable.
View return policy
Description
Gephyrin/GPHN Polyclonal antibody specifically detects Gephyrin/GPHN in Human samples. It is validated for Western Blot,Immunoprecipitation (Negative)
Specifications
| Gephyrin/GPHN | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation (Negative) | |
| CoraFluor 1 | |
| gephyrin, GPHRYN, KIAA1385GPHGEPH | |
| The immunogen this antibody was made to, maps to a region between residue 686 to 736 of human Gephyrin using the numbering given in entry NP_001019389.1 (GeneID 10243). | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience | |
| 10243 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction