missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gephyrin/GPHN Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-34859DL594
This item is not returnable.
View return policy
Description
Gephyrin/GPHN Polyclonal antibody specifically detects Gephyrin/GPHN in Human samples. It is validated for Western Blot
Specifications
| Gephyrin/GPHN | |
| DyLight 594 | |
| Cell Biology, Cytoskeleton Markers, Neuroscience | |
| Store at 4°C in the dark. | |
| IgG |
| Polyclonal | |
| 0.1 mL | |
| Primary | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction