missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gephyrin Recombinant Rabbit Monoclonal Antibody (9D0B0)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535565
This item is not returnable.
View return policy
Description
Immunogen sequence: MATEGMILTN HDHQIRVGVL TVSDSCFRNL AEDRSGINLK DLVQDPSLLG GTISAYKIVP DEIEEIKETL IDWCDEKELN LILTTGGTGF APRDVTPEAT.
Gephyrin is a neuronal assembly protein that anchors inhibitory neurotransmitter receptors to the postsynaptic cytoskeleton via high affinity binding to a receptor subunit domain and tubulin dimers. In nonneuronal tissues, the encoded protein is also required for molybdenum cofactor biosynthesis. Mutations in this gene may be associated with the neurological condition hyperplexia and also lead to molybdenum cofactor deficiency. Numerous alternatively spliced transcript variants encoding different isoforms have been described; however, the full-length nature of all transcript variants is not currently known.
Specifications
| Gephyrin | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q03555, Q8BUV3, Q9NQX3 | |
| Gphn | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Gephyrin (NP_001019389). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 9D0B0 | |
| Unconjugated | |
| Gphn | |
| 5730552E08Rik; AI662856; BC027112; C230040D23; Domain E; Domain G; EGK_18299; Geph; gephyrin; gephyrin isoform; Gph; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase; Putative glycine receptor-tubulin linker protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10243, 268566, 64845 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction