missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gephyrin Recombinant Rabbit Monoclonal Antibody (23GB5590), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 186.00 - € 446.00
Specifications
| Antigen | Gephyrin |
|---|---|
| Clone | 23GB5590 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087562
![]() |
Invitrogen™
MA552882 |
100 μL |
€ 446.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087858
|
Invitrogen™
MA552883 |
20 μL |
€ 186.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gephyrin Recombinant Monoclonal antibody specifically detects Gephyrin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Gephyrin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q03555, Q8BUV3, Q9NQX3 | |
| 10243, 268566, 64845 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB5590 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 5730552E08Rik; AI662856; BC027112; C230040D23; Domain E; Domain G; EGK_18299; Geph; gephyrin; gephyrin isoform; Gph; GPHN; GPHRYN; HKPX1; KIAA1385; MOCODC; Molybdopterin adenylyltransferase; Molybdopterin molybdenumtransferase; MPT adenylyltransferase; MPT Mo-transferase; Putative glycine receptor-tubulin linker protein | |
| A synthesized peptide derived from human Gephyrin (1-54AA). | |
| Primary | |
| -20°C | |
| Gphn |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title