Learn More
GFAP Antibody (CL2713) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76477AF488
Product Code. 30289257
  • € 748.00 / 0.10mL
Description
GFAP Monoclonal antibody specifically detects GFAP in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GFAP
Monoclonal
Alexa Fluor 488
ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
This antibody was developed using a recombinant protein derived from P14136, with the exact immunogen sequence remaining proprietary.
0.1 mL
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
2670
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL2713
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat