Learn More
GFAP Antibody (GFAP/8616R), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24136C
Code produit. 18528419
  • € 679.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
GFAP Monoclonal antibody specifically detects GFAP in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
GFAP
Monoclonal
DyLight 650
ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein
Recombinant fragment (around aa150-350) of human GFAP protein (exact sequence is proprietary)
0.1 mL
Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers
2670
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
GFAP/8616R