missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Antibody (rGFAP/8685), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24332DL594
This item is not returnable.
View return policy
Description
GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GFAP | |
| Monoclonal | |
| DyLight 594 | |
| ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
| Recombinant fragment (around aa150-350) of human GFAP protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 2670 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rGFAP/8685 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction