missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | GFAP |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | rGFAP/8685 |
| Conjugate | mFluor Violet 450 SE |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein |
| Host Species | Mouse |
| Immunogen | Recombinant fragment (around aa150-350) of human GFAP protein (exact sequence is proprietary) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?