missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Antibody (rGFAP/8685), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23834-100ug
This item is not returnable.
View return policy
Description
GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GFAP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 0.25-2 μg/mL | |
| ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
| Recombinant fragment (around aa150-350) of human GFAP protein (exact sequence is proprietary) | |
| 100 μg | |
| Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 2670 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rGFAP/8685 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction