missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Antibody (rGFAP/9150), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24050MFV500
This item is not returnable.
View return policy
Description
GFAP Monoclonal antibody specifically detects GFAP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GFAP | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
| Recombinant fragment (around aa100-300) of human GFAP protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 2670 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| rGFAP/9150 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction