missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Antibody (rGFAP/9150), Novus Biologicals™
Mouse Monoclonal Antibody
€ 232.00 - € 462.00
Specifications
| Antigen | GFAP |
|---|---|
| Clone | rGFAP/9150 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 0.25-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629044
|
Novus Biologicals
NBP3-23853-20ug |
20 μg |
€ 232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18619324
|
Novus Biologicals
NBP3-23853-100ug |
100 μg |
€ 462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GFAP Monoclonal antibody specifically detects GFAP in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| GFAP | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 2670 | |
| IgG2b κ | |
| Protein A or G purified |
| rGFAP/9150 | |
| Immunohistochemistry-Paraffin : 0.25-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
| Recombinant fragment (around aa100-300) of human GFAP protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title