missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Antibody (SPM248) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34401-0.1mg
This item is not returnable.
View return policy
Description
GFAP Monoclonal antibody specifically detects GFAP in Human, Mouse, Rat, Porcine, Bovine, Chicken, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| GFAP | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.5 - 1 μg/mL, Flow Cytometry 0.5 - 1 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1 - 2 μg/mL, Immunohistochemistry-Paraffin 0.5 - 1 μg/mL, CyTOF-ready | |
| ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
| GFAP isolated from pig spinal cord (Uniprot: P14136) | |
| 0.1 mg | |
| Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 2670 | |
| Store at -20°C to -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| SPM248 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Chicken, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction