missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Antibody (SPM248) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-34401JF525
This item is not returnable.
View return policy
Description
GFAP Monoclonal antibody specifically detects GFAP in Bovine,Chicken,Human,Mouse,Porcine,Rabbit,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),CyTOF,Immunocytochemistry/ Immunofluorescence
Specifications
| GFAP | |
| Monoclonal | |
| Janelia Fluor 525 | |
| ALXDRD, FLJ45472, GFAP, GFAP astrocytes, glial fibrillary acidic protein | |
| GFAP isolated from pig spinal cord (Uniprot: P14136) | |
| 0.1 mL | |
| Astrocyte Markers, Cancer, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Inflammation, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 2670 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), CyTOF, Immunocytochemistry/Immunofluorescence | |
| SPM248 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Chicken, Human, Mouse, Pig, Rabbit, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction