missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFAP Monoclonal Antibody (OTI1A7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814532S
This item is not returnable.
View return policy
Description
GFAP Monoclonal antibody specifically detects GFAP in Human samples. It is validated for ELISASpecifications
| GFAP | |
| Monoclonal | |
| See Label | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16552 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3605 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| ELISA | |
| OTI1A7 | |
| Unconjugated | |
| IL17A | |
| 6330404F12Rik; 65 kDa glutamic acid decarboxylase; AI836096; ALXDRD; Astrocyte or Intermediate Filament Protein; cb345; class III intermediate filament protein; etID36982.3; FLJ45472; GAD(65); Gad2; Gad-2; GAD65; GAD-65; GFAP; GFAP epsilon; gfap protein; gfapl; glial fibrillary acidic protein; Glial Fibrillary Acidic Protein (GFAP); glial fibrillary acidic protein alpha; glutamate decarboxylase 2; Glutamate decarboxylase 65 kDa isoform; glutamic acid decarboxylase 2; I79_011607; intermediate filament; intermediate filament protein; intermediate filament protein; GFAP; Unknown (protein for MGC:139638); wu:fb34h11; wu:fk42c12; xx:af506734; zgc:110485; zGFAP; intermediate filament protein; zrf-1 antigen | |
| Full length human recombinant protein of human IL-17A (NP_002181) produced in 293 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction