Learn More
GFAP Mouse anti-Human, Pig, Clone: GA-5, Abnova™
Mouse monoclonal antibody raised against human GFAP.
Brand: Abnova MAB12623.50ug
Description
This gene encodes one of the major intermediate filament proteins of mature astrocytes. It is used as a marker to distinguish astrocytes from other glial cells during development. Mutations in this gene cause Alexander disease, a rare disorder of astrocytes in the central nervous system. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Specifications
| GFAP | |
| Monoclonal | |
| Unconjugated | |
| Immunocytochemistry (1-4 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| FLJ45472 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 2670 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| GA-5 | |
| Mouse monoclonal antibody raised against human GFAP. | |
| In PBS, pH 7.4 (0.09% sodium azide). | |
| GFAP | |
| Pig spinal cord | |
| 50 μg | |
| Primary | |
| Human, Pig | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.