missing translation for 'onlineSavingsMsg'
Learn More

GFP Antibody (454505), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals FAB42401B

Product Code. 18638613

  • € 688.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GFP Monoclonal antibody specifically detects GFP in Broad Species Reactivity samples. It is validated for Immunoprecipitation, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GFP
Monoclonal
Biotin
PBS
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Store at 4°C in the dark.
IgG1
Immunoprecipitation, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
454505
Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
E. coli-derived recombinant GFPuv, Ser2-Lys238, Accession # P42212
0.1 mL
Epitope Tags
Broad Species Reactivity
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.