missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody (454505), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB42401E
This item is not returnable.
View return policy
Description
GFP Monoclonal antibody specifically detects GFP in Broad Species Reactivity samples. It is validated for Immunoprecipitation, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Specifications
| GFP | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunoprecipitation, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 454505 | |
| Immunoprecipitation, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| E. coli-derived recombinant GFPuv, Ser2-Lys238, Accession # P42212 | |
| 0.1 mL | |
| Epitope Tags | |
| Broad Species Reactivity | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction