missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody (454505R), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB42401RCL1
This item is not returnable.
View return policy
Description
GFP Monoclonal antibody specifically detects GFP in Broad Species Reactivity samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
| GFP | |
| Monoclonal | |
| CoraFluor 1 | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| E.coli-derived recombinant GFPuv, Ser2-Lys238, Accession # P42212 | |
| 0.1 mL | |
| Epitope Tags | |
| Broad Species Reactivity | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 454505R | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction