missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody (454505R), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB42401RMFV450
This item is not returnable.
View return policy
Description
GFP Monoclonal antibody specifically detects GFP in Broad Species Reactivity samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
| GFP | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Epitope Tags | |
| Broad Species Reactivity | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| 454505R | |
| Flow Cytometry, Immunocytochemistry | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| E.coli-derived recombinant GFPuv, Ser2-Lys238, Accession # P42212 | |
| RUO | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction