Learn More
GFP Antibody (4B10B2), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-22111AF647
Product Code. 18169514
  • € 579.00 / 0.10mL
Description
GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GFP
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Ammonium sulfate precipitation
RUO
Non-species specific
Purified
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
4B10B2
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp)
Purified recombinant fragment of GFP expressed in E. coli.
0.1 mL
Primary
Store at 4C in the dark.
IgG2a