missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-50059AF532
This item is not returnable.
View return policy
Description
GFP Polyclonal antibody specifically detects GFP in Nonspecies specific samples. It is validated for Western Blot, Immunofluorescence
Specifications
| GFP | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Recombinant AcGFP expressed in and purified from E. coli. (UniProt: Q6YGZ0) | |
| 0.1 mL | |
| Epitope Tags | |
| Non-species specific | |
| Purified |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction