Learn More
GFP Antibody, Novus Biologicals™
Product Code. 18750264 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mg
1mg
25µg
Quantity:
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize Quantity
18750264 25µg -
18730254 0.1mg -
30156535 1mg 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18750264 Supplier Novus Biologicals Supplier No. NB600308SS
Rabbit Polyclonal Antibody
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Electron Microscopy, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Gene Symbols GFP
Purification Method Affinity Purified
Target Species Non-species specific, C. elegans, Plant