missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, PE/Cy5.5, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-308PECY55
This item is not returnable.
View return policy
Description
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Flow Cytometry, Knockdown Validated.
Specifications
| GFP | |
| Polyclonal | |
| Flow Cytometry, KnockDown Validated | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Rabbit | |
| Affinity Purified | |
| Biologically Active Proteins, Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
| No reaction was observed against Human, Mouse or Rat serum proteins. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry, KnockDown | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| GFP | |
| The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. | |
| 0.1 mL | |
| Primary | |
| Non-species specific, C. elegans, Plant | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction