Learn More
IBA Lifesciences GFP-Strep-tag control protein
GFP-Strep-tag control protein for Strep-Tactin™ and Strep-Tactin™XT applications
Brand:  IBA Lifesciences 2-1006-005
Additional Details : Weight : 0.00050kg
 View more versions of this product
Product Code. 15593067
  • € 120.00 / 500µg
Estimated Shipment: 15-04-2024
to see stock.
Add to basket
Description
Description
  • GFP control protein consisting of the green fluorescent protein (GFP) C-terminally tagged with the Strep-tag™II or Twin-Strep-tag™.
  • It is suitable for test purifications, detections, and immobilizations with Strep-Tactin™ or Strep-Tactin™XT.  
Specifications
Specifications
5 mL
500 μg of a 30 kDa protein
Strep-tag™II