missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFPT1 Antibody (SR2221), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22535-100ul
This item is not returnable.
View return policy
Description
GFPT1 Monoclonal antibody specifically detects GFPT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
Specifications
| GFPT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| SR2221 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50 | |
| glucosamine--fructose-6-phosphate aminotransferase [isomerizing] 1, CMSTA1, GFA, GFAT, GFAT 1, GFAT1, GFAT1m, GFPT, GFPT1L, glutamine--fructose-6-phosphate transaminase 1, MSLG | |
| A synthesized peptide derived from human GFPT1 (Uniprot #: Q06210) | |
| 100 μg | |
| metabolism | |
| 2673 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction