missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFPT1 Mouse anti-Human, Monkey, Rat, Clone:1F1A4, Abnova™
Mouse monoclonal antibody raised against recombinant human GFPT1.
Brand: Abnova MAB17183.100ug
This item is not returnable.
View return policy
Description
This gene encodes the first and rate-limiting enzyme of the hexosamine pathway and controls the flux of glucose into the hexosamine pathway. The product of this gene catalyzes the formation of glucosamine 6-phosphate. [provided by RefSeq]Specifications
| GFPT1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry (1:200-1:1000) Flow Cytometry (1:200-1:400) The optimal working dilution should be determined by the end user. |
|
| GFPT1 | |
| Mouse | |
| 100 μg | |
| Human, Monkey, Rat | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
| 1F1A4 | |
| glutamine-fructose-6-phosphate transaminase 1 | |
| In PBS (0.05% sodium azide) | |
| GFA, GFAT, GFAT1, GFAT1m, GFPT | |
| Recombinant protein corresponding to amino acid 536-681 of human GFPT1 from E. coli. | |
| 2673 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction