missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGA2 Antibody (OTI1D2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71725B
This item is not returnable.
View return policy
Description
GGA2 Monoclonal specifically detects GGA2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GGA2 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| GGA2 | |
| Full length recombinant protein of human GGA2 (NP_055859) produced in HEK293T cells | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot | |
| OTI1D2 | |
| Western Blot | |
| FLJ20966, Gamma-adaptin-related protein 2, golgi associated, gamma adaptin ear containing, ARF binding protein 2, golgi-associated, gamma adaptin ear containing, ARF binding protein 2, Golgi-localized, gamma ear-containing, ARF-binding protein 2, KIAA1080ADP-ribosylation factor-binding protein GGA2, VEAR, VHS domain and ear domain of gamma-adaptin, VHS domain and ear domain-containing protein | |
| Mouse | |
| Affinity Purified | |
| Golgi Apparatus Markers, Membrane Trafficking and Chaperones | |
| 23062.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction