missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GGCX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553770
This item is not returnable.
View return policy
Description
Immunogen sequence: FPGLHLENFV SEDLGNTSIQ LLQGEVTVEL VAEQKNQTLR EGEKMQLPAG EYHKVYTTSP SPSCYMYVYV NTTELALEQD LAYLQELKEK VENGSETGPL PPELQPLLEG EVK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.
GGCX (gamma-glutamyl carboxylase), also known as GC or VKCFD1 (Vitamin K-dependent gamma-carboxylase), is a 758 amino acid multi-pass membrane protein. Localized to the membrane of the endoplasmic reticulum, GGCX functions to mediate the vitamin K-dependent carboxylation of glutamate residues on target proteins, thereby producing calcium binding gamma-carboxyglutamate (Gla) residues on these proteins and simultaneously converting vitamin K to vitamin K epoxide. GGCX exists as a monomer and, via its ability to modify glutamate residues, it accomplishes the post-translational changes that are necessary for the activity of all vitamin K-dependent proteins (such as blood coagulation and bone matrix proteins). Defects in the gene encoding GGCX are the cause of combined deficiency of vitamin K-dependent clotting factors 1 (VKCFD1) and PXE-like disorder with multiple coagulation factor deficiency, both of which are characterized by abnormal skin, blood or bone function.
Specifications
| GGCX | |
| Polyclonal | |
| Unconjugated | |
| Ggcx | |
| FLJ26629; Gamma-glutamyl carboxylase; GC; GGCX; Peptidyl-glutamate 4-carboxylase; vitamin K gamma glutamyl carboxylase; Vitamin K-dependent gamma-carboxylase; VKCFD1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2677 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P38435 | |
| Ggcx | |
| Recombinant protein corresponding to Human GGCX. Recombinant protein control fragment (Product #RP-91226). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction