missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GGN Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145183
This item is not returnable.
View return policy
Description
Immunogen sequence: SGAISYAEVL KQGPLPPGAA RPLGEVSRGA QEAEGGDGDG EGCSGPPSAP ASQARALPPP PYTTFPGSKP KFDWVSAPDG PERHFRFNGA G Antigen sequence identity: Mouse 78%; Rat 86%.
May be involved in spermatogenesis.
Specifications
| GGN | |
| Polyclonal | |
| Unconjugated | |
| GGN | |
| AI593290; Gametogenetin; GGN | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 199720 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86UU5 | |
| GGN | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 280-370. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction