Learn More
GGPS1 Mouse anti-Human, Clone: OTI3F9, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501731
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the prenyltransferase family and encodes a protein with geranylgeranyl diphosphate (GGPP) synthase activity. The enzyme catalyzes the synthesis of GGPP from farnesyl diphosphate and isopentenyl diphosphate. GGPP is an important molecule responsible for the C20-prenylation of proteins and for the regulation of a nuclear hormone receptor. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| GGPS1 | |
| Monoclonal | |
| Unconjugated | |
| GGPS1 | |
| GGPPS, GGPPS1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9453 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3F9 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O95749 | |
| GGPS1 | |
| Full length human recombit protein of human GGPS1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.