missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GGPS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528470
This item is not returnable.
View return policy
Description
Recommended positive controls: PC-3, SK-N-SH. Predicted reactivity: Mouse (94%), Rat (94%), Zebrafish (82%), Xenopus laevis (87%), Rhesus Monkey (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the prenyltransferase family and encodes a protein with geranylgeranyl diphosphate (GGPP) synthase activity. The enzyme catalyzes the synthesis of GGPP from farnesyl diphosphate and isopentenyl diphosphate. GGPP is an important molecule responsible for the C20-prenylation of proteins and for the regulation of a nuclear hormone receptor. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| GGPS1 | |
| Polyclonal | |
| Unconjugated | |
| GGPS1 | |
| (2E,6E)-farnesyl diphosphate synthase; 1810026C22Rik; 9530089B04Rik; AI843169; C79210; Crlf3; dimethylallyltranstransferase; Farnesyl diphosphate synthase; farnesyltranstransferase; Geranylgeranyl diphosphate synthase; geranylgeranyl diphosphate synthase 1; geranylgeranyl pyrophosphate synthase; geranylgeranyl pyrophosphate synthetase; geranyltranstransferase; GGPP synthase; GGPP synthetase; GGPPS; GGPPS1; GGPPSase; GGPS1; rGGPS1a; rGGPS1a1; rGGPS1a2; rGGPS1a3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9453 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.9 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O95749 | |
| GGPS1 | |
| Recombinant protein encompassing a sequence within the center region of human GGPS1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction