missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 359.00 - € 515.00
Specifications
| Antigen | GGT1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
GGT1 Polyclonal specifically detects GGT1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| GGT1 | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 2678 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MFNSSEQSQKGGLSVAVPGEIRGYELAHQRHG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| CD224, CD224 antigen, D22S672, D22S732, EC 2.3.2.2, gamma-glutamyl transpeptidase, gamma-glutamyltransferase 1MGC96904, gamma-glutamyltranspeptidase 1, GGT 1, GGTMGC96963, glutamyl transpeptidase, GTG, MGC96892 | |
| GGT1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title