missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GHITM Antibody (42), Novus Biologicals™
Mouse Monoclonal Antibody
€ 216.00 - € 359.00
Specifications
| Antigen | GHITM |
|---|---|
| Clone | 42 |
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18600267
|
Novus Biologicals
NBP3-12773-100ul |
100 μg |
€ 359.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660809
|
Novus Biologicals
NBP3-12773-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GHITM Monoclonal antibody specifically detects GHITM in Invertebrate samples. It is validated for ELISASpecifications
| GHITM | |
| ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 27069 | |
| IgG2a | |
| Protein A purified |
| 42 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Invertebrate | |
| Dermal papilla-derived protein 2, DERP2MICS1, DKFZp566C0746, FLJ26584, growth hormone inducible transmembrane protein, growth hormone-inducible transmembrane protein, HSPC282, Mitochondrial morphology and cristae structure 1, PTD010, TMBIM5My021, transmembrane BAX inhibitor motif containing 5, Transmembrane BAX inhibitor motif-containing protein 5 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dermatophagoides pteronyssinus GHITM (Uniprot#: P49278; Gln19-Asp146). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title