Learn More
GHITM Antibody (42), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-12773PCP
Product Code. 18654537
  • € 407.00 / 0.10mL
Description
GHITM Monoclonal antibody specifically detects GHITM in Invertebrate samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GHITM
Monoclonal
PerCP
PBS
Mouse
Protein A purified
RUO
27069
Store at 4C in the dark.
IgG2a
ELISA
42
ELISA
Dermal papilla-derived protein 2, DERP2MICS1, DKFZp566C0746, FLJ26584, growth hormone inducible transmembrane protein, growth hormone-inducible transmembrane protein, HSPC282, Mitochondrial morphology and cristae structure 1, PTD010, TMBIM5My021, transmembrane BAX inhibitor motif containing 5, Transmembrane BAX inhibitor motif-containing protein 5
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Dermatophagoides pteronyssinus GHITM (Uniprot#: P49278; Gln19-Asp146).
0.1 mL
Primary
Invertebrate