Learn More
Ghrelin Receptor/GHSR Antibody (502430), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8370W
Product Code. 18630370
  • € 622.00 / 0.10mL
Description
Ghrelin Receptor/GHSR Monoclonal antibody specifically detects Ghrelin Receptor/GHSR in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ghrelin Receptor/GHSR
Monoclonal
DyLight 650
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry, CyTOF
502430
Flow Cytometry, CyTOF-ready
GH-releasing peptide receptor, Ghrelin receptor, GHRP, GHS-R, growth hormone secretagogue receptor, growth hormone secretagogue receptor type 1
NS0 mouse myeloma cell line transfected with human GHSR, Accession # NP_940799
0.1 mL
GPCR
2693
Store at 4°C in the dark.