missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GHRHR Polyclonal Antibody

Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Elephant (94%) Panda, Bat, Horse, Pig (88%) Marmoset, Zebu, Bovine, Dog (81%).
Growth Hormone Releasing Hormone (GHRH) is a member of a family of peptide hormones that also includes VIP (vasoactive intestinal polypeptide) and PACAP (pituitary adenylate cyclase-activating polypeptide). It is a hypothalamic peptide that binds to G-protein coupled receptors to stimulate secretion and synthesis of growth hormone (GH). GHRH receptor proteins have been identified on the plasma membrane of primary human tumor cells, and are thought to be involved in the growth of a variety of cancer cells.
Specifications
Specifications
| Antigen | GHRHR |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | Ghrhr |
| Gene Accession No. | Q02643 |
| Gene Alias | GHRFR; GHRH receptor; GHRHR; Ghrh-r; GHRHREC; GHRHRpsv; GRF receptor; GRFR; Growth hormone - releasing receptor; growth hormone releasing hormone receptor; growth hormone-releasing factor receptor; Growth hormone-releasing hormone receptor; IGHD1B; lit; little |
| Gene Symbols | Ghrhr |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?