missing translation for 'onlineSavingsMsg'
Learn More

GI254023X, MedChemExpress

Product Code. 16491899
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491899 1 mg 1mg
16440448 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491899 Supplier Medchem Express Supplier No. HY19956

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GI254023X is a potent MMP9 and ADAM10 inhibitor with IC50s of 2.5 and 5.3 nM, respectively.

GI254023X is a potent MMP9 and ADAM10 inhibitor with IC50s of 2.5 and 5.3 nM, respectively.

Specifications

Chemical Name or Material GI254023X
Physical Form Solid
CAS 260264-93-5
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C21H33N3O4
For Use With (Application) Cancer-Kinase/protease
Synonym GI4023 SRI028594
Solubility Information DMSO : 100 mg/mL (255.43 mM; Need ultrasonic)
SMILES O=C(N[C@H](C(NC)=O)C(C)(C)C)[C@@H]([C@@H](N(C=O)O)C)CCCC1=CC=CC=C1
Molecular Weight (g/mol) 391.5
Formula Weight 391.5
Percent Purity 98.2%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.