missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Giardia lamblia 65 kDa Monoclonal Antibody (6292)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA547568

Product Code. 17856657

  • € 504.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specificity: Specific to the 65 kDa Giardia antigen. Detects cysts. Comments: Best ELISA pairs (capture/conjugate): Giardia lamblia 65 kDa Monoclonal antibody (Product # MA5-47567)/Giardia lamblia 65 kDa Monoclonal antibody (Product # MA5-47568). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

Giardiasisis a diarrhoeal illness caused by a single celled microscopic protozoan parasite, Giardia lamblia, also known as Giardia intestinalis. Giardia lamblia exists in two forms, an active form called a trophozoite, and an inactive form called a cyst. The active trophozoite attaches to the lining of the small intestine and is responsible for causing the signs and symptoms of giardiasis. The trophozoite cannot live long outside of the body and spread of infection is via the cyst which is excreted in the host's faeces. When it is ingested, stomach acid activates the cyst, and the cyst develops into the disease causing trophozoite in the new host. Giardiasis is diagnosed by finding cysts or trophozoites in the faeces.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Giardia lamblia 65 kDa
Monoclonal
100 μg/mL
PBS with 0.1% sodium azide; pH 7.2
Mouse
Protein A
RUO
Protozoa
Antibody
IgG2b
ELISA, Immunocytochemistry
6292
Unconjugated
G. lamblia
Proprietary sequence.
100 μg
Primary
4°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.