missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Giardia lamblia Antibody (BB1.1E5), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08870F
This item is not returnable.
View return policy
Description
Giardia lamblia Monoclonal antibody specifically detects Giardia lamblia in Protozoa samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Giardia lamblia | |
| Monoclonal | |
| FITC | |
| G lamblia; Giardia intestinalis | |
| Giardia lamblia | |
| 100 μg | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunofluorescence | |
| BB1.1E5 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Protozoa | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction