Learn More
Thermo Scientific Chemicals Giemsa Stain, High Purity Biological Stain
Catalog Number p-29912
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
51811-82-6
C14H14ClN3S
291.80
MFCD00012112,MFCD00081642
NALREUIWICQLPS-UHFFFAOYSA-N
13735
(7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
[Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1
51811-82-6
C14H14ClN3S
291.80
MFCD00012112,MFCD00081642
NALREUIWICQLPS-UHFFFAOYSA-N
13735
(7-aminophenothiazin-3-ylidene)-dimethylazanium;chloride
[Cl-].C[N+](C)=C1C=CC2=NC3=CC=C(N)C=C3SC2=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Absorbance 950 min. (in MeOH, on dried substance), 600 min. (in MeOH, on dried substance) A (1%/1cm)
Melting Point 300.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Infrared Spectrum Authentic
Lambda Max 640 to 650nm (in MeOH), 520 to 525nm (in MeOH)
Loss on Drying 10% max. (110°C)
Grade Pure
Packaging Glass bottle
Color Black to Green
Quantity 5 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Giemsa Stain
Show More Show Less
compliance-icons
Product Identifier
  • Giemsa Stain
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.